Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRB3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501733762 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-3762 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-3762 Supplier Proteintech Group Inc Supplier No. 221901AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PRB3 protein is identical to the gross cystic disease fluid protein GCDFP-15, secretory actin binding protein (SABP) and glycoprotein 17 (GP-17). GCDFP-15 is expressed by breast carcinomas (primary and metastatic lesions) with apocrine features.

Specifications

Antigen PRB3
Applications Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PRB3
Gene Accession No. Q04118
Gene Alias G1, PRB3, PRG, Proline rich protein G1
Gene Symbols PRB3, Prb3-ps
Host Species Rabbit
Immunogen PRB3 Fusion Protein Ag16760
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5544, 639802
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.