Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRB4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110212
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110212 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110212 Supplier Invitrogen™ Supplier No. PA5110212
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SPPGKPQGPP QQEGNKPQGP PPPGKPQGPP PAGGNPQQPQ APPAGKPQGP PPPPQGGRPP RPAQGQQPPQ.

This gene encodes a member of the heterogeneous family of basic, proline-rich, human salivary glycoproteins. The encoded preproprotein undergoes proteolytic processing to generate one or more mature peptides before secretion from the parotid glands. Multiple alleles of this gene exhibiting variations in the length of the tandem repeats have been identified. The reference genome encodes the Small allele. This gene is located in a cluster of closely related salivary proline-rich proteins on chromosome 12. Alternative splicing results in multiple transcript variants encoding different isoforms that may undergo similar proteolytic processing. [provided by RefSeq, Nov 2015].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRB4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PRB4
Gene Accession No. P10163
Gene Alias basic salivary proline-rich protein 4; BstNI type basic salivary proline-rich protein 4; parotid o protein; Peptide P-D; Po; PRB4; proline rich protein BstNI subfamily 4; proline-rich protein BstNI subfamily 4; salivary proline-rich protein II-1; salivary proline-rich protein Po
Gene Symbols PRB4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 178-247 of human PRB4 (NP_002714.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5545
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.