Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRDM12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599266
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599266 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599266 Supplier Invitrogen™ Supplier No. PA599266
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PRDM12.

This gene encodes a transcriptional regulator of sensory neuronal specification that plays a critical role in pain perception. The encoded protein contains an N-terminal PRDI-BF1 and RIZ homology (PR) domain, a SET domain, and three C-terminal C2H2 zinc finger DNA-binding domains. Naturally occurring mutations in this gene are associated with congenital insensitivity to pain (CIP), and hereditary sensory and autonomic neuropathies (HSAN's) affecting peripheral sensory and autonomic neurons. Deregulation of this gene is associated with solid cancers and hematological malignancies including chronic myeloid leukemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRDM12
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PRDM12
Gene Accession No. A2AJ77, Q9H4Q4
Gene Alias Gm998; HSAN8; PFM9; PR domain 12; PR domain containing 12; PR domain zinc finger protein 12; PR domain-containing protein 12; PRDM12; PR-domain containing protein 12; PR-domain zinc finger protein 12; RP11-57C19.7
Gene Symbols PRDM12
Host Species Rabbit
Immunogen A synthesized peptide derived from human PRDM12(Accession Q9H4Q4), corresponding to amino acid residues L198-C248.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 381359, 59335
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.