Learn More
Invitrogen™ PRDM12 Polyclonal Antibody

Description
Immunogen sequence: LGIFSKTWIK AGTEMGPFTG RVIAPEHVDI CKNNNLMWEV FNEDGTVRYF IDASQEDHRS WMTYIKCARN EQEQNLEVVQ IGTSIFYKAI EMIPPDQELL VWYGNSHNTF LGIPGVPGLE EDQKKNKHED FHPADSAAGP AGRMRCVICH RGFNSRSNLR SHMRIHTLDK PFVCRFCNRR FSQSSTLRNH VRLHTGERPY KCQVCQSAYS QLAGLRAHQK SARHRPPSTA L.
Specifications
Specifications
| Antigen | PRDM12 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.95 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | PRDM12 |
| Gene Accession No. | A2AJ77, Q9H4Q4 |
| Gene Alias | Gm998; HSAN8; PFM9; PR domain 12; PR domain containing 12; PR domain zinc finger protein 12; PR domain-containing protein 12; PRDM12; PR-domain containing protein 12; PR-domain zinc finger protein 12; RP11-57C19.7 |
| Gene Symbols | PRDM12 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.