Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRDM2 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501735738 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5738 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5738 Supplier Proteintech Group Inc Supplier No. 277181AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PRDM2 is a 250-kD nuclear protein containing 8 zinc finger motifs. It contains an RB-binding motif that is related to that of the adenovirus E1A oncoprotein; RIZ also shares an antigenic epitope with the C terminus of E1A. It is expressed in human retinoblastoma cells and at low levels in all other human cell lines examined. Using fluorescence in situ hybridization, the PRDM2 gene has been mapped to 1p36. PRDM2 may be a candidate target of 1p36 alterations that commonly occur in neuroendocrine, breast, liver, colon, skin, and lymphoid tumors.

Specifications

Antigen PRDM2
Applications Western Blot
Classification Polyclonal
Concentration 0.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PRDM2
Gene Accession No. Q13029
Gene Alias GATA 3 binding protein G3B, HUMHOXY1, KMT8, Lysine N methyltransferase 8, MTB ZF, MTE binding protein, PR domain containing protein 2, PRDM2, RIZ, RIZ1, RIZ2, Zinc finger protein RIZ
Gene Symbols PRDM2
Host Species Rabbit
Immunogen PRDM2 Fusion Protein Ag26769
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7799
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.