Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRDM5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01675748 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-748 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-748 Supplier Invitrogen™ Supplier No. PA141182
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Suggested positive control: HL-60.

PRDM5, a novel member of PR domain protein family, contains a PR-domain at the -NH2 terminus followed by 16 zinc finger motifs. It is found to have a role in cell differentiation and tumorigenesis. Reports suggest PRDM5 causes G2/M arrest and apoptosis upon infection of tumor cells and has been found to be silenced in breast, ovarian and lung cancers, suggesting a potential clinical significance.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRDM5
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% gelatin and 0.05% sodium azide
Gene PRDM5
Gene Accession No. Q9NQX1
Gene Alias 4432417F03Rik; 6530401I24Rik; AI197291; BCS2; E130112L17Rik; PFM2; PR domain 5; PR domain containing 5; PR domain zinc finger protein 5; PR domain-containing protein 5; PR/SET domain 5; PRDM5; unm hi61; un-named hi61
Gene Symbols PRDM5
Host Species Rabbit
Immunogen A portion of amino acids 100-200 of human PRDM5 protein.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11107
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.