Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRDM8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107223
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107223 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107223 Supplier Invitrogen™ Supplier No. PA5107223
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

Probable histone methyltransferase, preferentially acting on 'Lys-9' of histone H3. Involved in the control of steroidogenesis through transcriptional repression of steroidogenesis marker genes such as CYP17A1 and LHCGR. Forms with BHLHE22 a transcriptional repressor complex controlling genes involved in neural development and neuronal differentiation. In the retina, it is required for rod bipolar and type 2 OFF-cone bipolar cell survival.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRDM8
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene PRDM8
Gene Accession No. Q9NQV8
Gene Alias EPM10; PFM5; PR domain 8; PR domain zinc finger protein 8; PR domain-containing protein 8; PRDM8; PR-domain containing protein 8
Gene Symbols PRDM8
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human PRDM8.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56978
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.