Learn More
Invitrogen™ PRDX4 Polyclonal Antibody

Description
PA3-753 detects Peroxiredoxin 4 protein in human samples. This antibody is specific for Prx 4 and shows no cross-reactivity with other Prx isoforms. PA3-753 has successfully been used in Western blot and immunocytochemistry procedures. By Western blot, this antibody detects an ~23 kDa and ~20 kDa protein representing pro-Prx 4 and mature Prx 4, respectively, in human PC3 cell lysate. The PA3-753 antigen is purified human recombinant Prx 4 from K562 cells.
Specifications
Specifications
| Antigen | PRDX4 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | whole serum with 0.05% sodium azide |
| Gene | PRDX4 |
| Gene Accession No. | Q13162 |
| Gene Alias | Antioxidant enzyme AOE372; AOE372; AOE37-2; EC 1.11.1.15; epididymis secretory sperm binding protein Li 97n; HEL-S-97n; peroxiredoxin 4; peroxiredoxin IV; Peroxiredoxin-4; Prdx4; Prx 4; Prx IV; Prx4; PRX-4; prx-IV; Rno2CysPrx04; Thioredoxin dependent peroxide reductase A0372; thioredoxin peroxidase (antioxidant enzyme); thioredoxin peroxidase AO372; Thioredoxin-dependent peroxide reductase A0372; TRANK |
| Gene Symbols | PRDX4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.