Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRDX5 Recombinant Rabbit Monoclonal Antibody (125)
GREENER_CHOICE

Catalog No. PIMA530782
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530782 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530782 Supplier Invitrogen™ Supplier No. MA530782
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human Peroxiredoxin 5/PRDX5.

Peroxiredoxin (Prx) is a growing peroxidase family, whose mammalian members have been known to connect with cell proliferation, differentiation, and apoptosis. Many isoforms (about 50 proteins), collected in accordance to the amino acid sequence homology, containing active site cysteine residue, and the thiol-specific antioxidant activity, distribute throughout all the kingdoms. Among them, mammalian Prx consists of 6 different members grouped into typical 2-Cys, atypical 2-Cys Prx, and 1-Cys Prx. Except Prx VI belonging to 1-Cys Prx subgroup, the other five 2-Cys Prx isotypes have the thioredoxin-dependent peroxidase (TPx) activity utilizing thioredoxin, thioredoxin reductase, and NADPH as a reducing system. Mammalian Prxs are 20-30 kilodalton in molecular size and vary in subcellular localization: Prx I, II, and VI in cytosol, Prx III in mitochondria, Prx IV in ER and secretion, Prx V showing complicated distribution including peroxisome, mitochondria and cytosol.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRDX5
Applications ELISA
Classification Recombinant Monoclonal
Clone 125
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene PRDX5
Gene Accession No. P30044
Gene Alias ACR1; Alu corepressor 1; Alu co-repressor 1; Antioxidant enzyme B166; AOEB166; AOPP; B166; epididymis secretory protein Li 55; HEL-S-55; Liver tissue 2D-page spot 2D-0014IV; liver tissue 2D-page spot 71B; peroxiredoxin 5; peroxiredoxin 6; peroxiredoxin V; Peroxiredoxin-5, mitochondrial; peroxisomal antioxidant enzyme; peroxisomal membrane protein 20; PLP; Pmp20; PRDX5; Prdx6; Prx5; PrxV; prx-V; SBBI10; Thioredoxin peroxidase PMP20; thioredoxin reductase; TPx type VI
Gene Symbols PRDX5
Host Species Rabbit
Immunogen Recombinant Human Peroxiredoxin 5/PRDX5 Protein (Met53-Leu214).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25824
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.