Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynamic Diagnostics Pre-filled 10% Neutral Buffered Formalin (NBF) Specimen Containers

Catalog No. 22899689
Change view
Click to view available options
Capacity:
20 mL Total
40 mL Total
60 mL Total
90 mL Total
Includes:
Pre-filled with 10 mL 10% NBF
Pre-filled with 20 mL 10% NBF
Pre-filled with 40 mL 10% NBF
Pre-filled with 60 mL 10% NBF
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity Includes
22-899-689 40 mL Total Pre-filled with 20 mL 10% NBF
22-899-688 20 mL Total Pre-filled with 10 mL 10% NBF
22-899-690 60 mL Total Pre-filled with 40 mL 10% NBF
22-899-691 90 mL Total Pre-filled with 60 mL 10% NBF
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-899-689 Supplier Dynamic Diagnostics Supplier No. 10151
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled

These Pre-filled 10% NBF Specimen Containers are well suited for large throughput laboratories and require a relatively short period of fixation, but can also be used for the long-term storage of tissue. Neutral buffered formalin (NBF) is the most commonly used fixative throughout the world for light microscopy.

  • Pre-filled with 10% neutral buffered formalin
  • Packaged in dispenser boxes with perforated tear-away lids to easily access bottles
  • Each bottle labeled for clinical use

Specifications

Capacity 40 mL Total
Concentration 6% Formaldehyde, 4% Alcohol
Grade Professional Grade
Product Type Formalin
Quantity 24 Containers/HistoPak Box, 4 HistoPak Boxes/Cs.
Solution Type Formalin
For Use With (Application) Tissue Fixation & Specimen Transport
Includes Pre-filled with 20 mL 10% NBF
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.