Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Pre-Filter for PA700 Cartridge Series

Catalog No. 17378716
Change view
Click to view available options
Pre-Filter for PA700 Cartridge Series
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
17-378-716 Pre-Filter for PA700 Cartridge Series
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-378-716 Supplier Honeywell™ Supplier No. PA7PR
This item is not returnable. View return policy

Pre-filter for PA700 cartridge series

  • Pre-filter for PA700 cartridge series
  • Compatible with the PA7HE filter cartridge
TRUSTED_SUSTAINABILITY

Specifications

Color White
Product Type Prefilter
For Use With (Equipment) Compatible with the PA7HE filter cartridge. (Not compatible with the PA7OVHE, PA7OVAGHE and PA7DEHE )
Latex Free Yes
Model PA7PR prefilter
Quantity 10 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.