Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pre-miR™ miRNA Precursor Negative Control #1

Catalog No. AM17110
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 nmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM17110 5 nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM17110 Supplier Invitrogen™ Supplier No. AM17110
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Pre-miR™ miRNA Precursor Negative Control #1

Pre-miR™ miRNA Precursors are designed to mimic endogenous miRNAs. Pre-miR miRNA Precursor Negative Control #1 is designed to use as a negative control for experiments using Pre-miR miRNA Precursors. Transfect Pre-miR miRNA Precursor Negative Control #1 using the same methodology as for your experimental Pre-miR miRNA Precursors, then use the target gene expression from the sample transfected with the Pre-miR miRNA Precursor Negative Control #1 as a baseline for evaluation of the effect of the experimental Pre-miR miRNA Precursor on target gene expression. Advantages of the Pre-miR™ miRNA Precursor Negative Control #1:

  • Optimized design results in correct strand utilization by RISC
  • Purified and ready to transfect

    About Pre-miR miRNA Precursors
    Pre-miR™ miRNA Precursor Molecules are small, chemically modified, double-stranded RNA molecules designed to mimic endogenous mature miRNAs. This ready-to-use miRNA mimic can be introduced into cells using transfection or electroporation parameters similar to those used for siRNAs. They enable detailed study of miRNA biological effects via gain-of-function experiments.

    Using Pre-miR miRNA Precursor Negative Control #1
    Pre-miR™ Negative Control #1 is a random sequence Pre-miR™ molecule that has been extensively tested in human cell lines and tissues and validated to produce no identifiable effects on known miRNA function. RNA oligonucleotides are susceptible to degradation by exogenous ribonucleases introduced during handling. Wear gloves when handling this product. Use RNase-free reagents, tubes, and barrier pipette tips. Upon receipt, store in a non-frost-free freezer at or below -20°C (dried oligonucleotides are shipped at ambient temperature).

    NOTE: Pre-miR™ miRNA Precursors are not hairpin constructs and should not be confused with pre-miRNAs.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Pre-miR™ miRNA is provided dried down in a single tube, along with Nuclease-free Water for resuspension, and should be stored at –20°C. It is guaranteed for 1 year when stored dried at –20°C.
    Format Tube
    Control Type Negative Control
    RNAi Type miRNA Mimic
    For Use With (Application) miRNA & Non-Coding RNA Analysis
    Label or Dye Unconjugated
    Product Line Ambion, Pre-miR
    Product Type miRNA Mimic
    Quantity 5 nmol
    Shipping Condition Room Temperature
    Where can I find more information about published miRNA sequences and annotation?

    Please visit miRBase: The microRNA database (http://www.mirbase.org/).

    I would like to order a mimic or inhibitor with a 5' label. Can you do this?

    We can label our Pre-miR and Anti-miR miRNA, but do not offer labeled mirVana Mimics and inhibitors. 5' modifications include Alexa Fluor 555, Alexa Fluor 647, FAM, and Cy3. Request a quote here (http://www.thermofisher.com/us/en/home/global/forms/custom-sirna-mirna-quote-request.html).

    The miRNA sequence I need to target is not in the miRBase database. Can I order a custom mimic or inhibitor?

    Yes, submit your unpublished miRNA sequences and/or order custom modified miRNA here (http://www.thermofisher.com/order/custom-genomic-products/tools/mirna/).

    What is the difference between Invitrogen Anti-miR inhibitors & Invitrogen Pre-miR miRNA Precursors and mirVana miRNA mimics and inhibitors?

    Invitrogen Anti-miR inhibitors & Invitrogen Pre-miR miRNA Precursors are our first generation mimic and inhibitors. Invitrogen Anti-miR miRNA Inhibitors are chemically modified, single stranded nucleic acids designed to specifically bind and inhibit endogenous microRNA (miRNA) molecules. These are ready to use inhibitors that can be introduced into cells using transfection or electroporation. Pre-designed inhibitors have been created for all miRNAs listed in miRBase version 15.0.
    Invitrogen Pre-miR miRNA Precursors are small, chemically modified double-stranded RNA molecules designed to mimic endogenous mature miRNAs. These ready-to-use miRNA mimics, which are similar to, but not identical to siRNAs, can be introduced into cells using transfection or electroporation parameters similar to those used for siRNAs. Please note, Pre-miR miRNA Precursors are not hairpin constructs and should not be confused with pre-miRNAs.
    mirVana miRNA mimics and inhibitors are our second generation of mimics and inhibitors. mirVana miRNA mimics are small, chemically double-stranded RNAs that mimic endogenous mature miRNAs and enable miRNA functional analysis by up-regulation of miRNA activity. They have enhanced specificity over the first generation mimics, are delivered as HPLC purified, and ready to transfect.
    miRNA inhibitors are small, chemically modified single-stranded RNA molecules designed to specifically bind to and inhibit endogenous miRNA molecules and enable miRNA functional analysis by down-regulation of miRNA activity. They have enhanced potency over our first generation inhibitors. Pre-designed inhibitors were created for all miRNAs listed in miRBase version 19.0. They come delivered as HPLC purified, and ready to transfect. Learn more about mirVana miRNA mimics and inhibitors through this video (https://www.youtube.com/watch?feature=player_embedded&v=Q3MO1lmT0kw). Both mimics and inhibitors are available individually or as libraries.
    They are compatible with both in vitro and in vivo applications, and have been validated with Lipofectamine RNAiMAX Transfection Reagent for use in cell-based systems, and with Invivofectamine 3.0 Transfection Reagent for in vivo delivery. These oligonucleotides are nontoxic and do not induce an immune response in the animal models tested. In vivo-ready mirVana miRNA mimics and inhibitors have been purified by HPLC and dialysis and are ready for immediate in vivo use. For all library inquiries, please email: rnailibraries@lifetech.com.

    What is the difference between siRNA and miRNA?

    siRNA mediates degradation of a target mRNA molecule in a sequence-specific manner, while miRNA suppresses translation without cleaving mRNA.

    What is miRNA?

    miRNA (microRNAs) are short, highly conserved small noncoding RNA molecules naturally occurring in the genomes of plants and animals. miRNAs are single stranded, 18-25 nucleotides long and regulate posttranscriptional mRNA expression, typically by binding to the 3' untranslated region (3'-UTR) of the complementary mRNA sequence, resulting in translational repression and gene silencing. Studies have shown that thousands of human protein-coding genes are regulated by miRNAs, indicating that miRNAs are “master regulators” of many important biological processes.


    WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.