Learn More
Thermo Scientific™ Pre-supplemented Dichloran Rose-bengal Chloramphenicol (DRBC) (ISO) Agar (Dehydrated)
Shop All Thermo Scientific ProductsDescription
Isolate and enumerate yeasts and molds associated with food spoilage with Thermo Scientific™ Oxoid™ Pre-supplemented Dichloran Rose-Bengal Chloramphenicol (DRBC) (ISO) Agar Base (Dehydrated) while adhering to ISO 21527-1: 2008. Due to the presence of antibiotic Chloramphenicol in the formulation, the medium provides selectivity against bacteria as well as reducing harmful chemical handling in the laboratory.
- Adheres to ISO 21527-1: 2008 standards.
- Selective for viable yeast and mold samples in food samples.
The inclusion of chloramphenicol in this medium suppresses bacterial growth, allowing molds, and yeasts to grow. The inhibition of spreading molds and the restriction of their colony size results in improved enumeration and detection. The reduced pH of DRBC (ISO) Agar increases the inhibition of yeasts by the rose-bengal. Pre-supplemented DRBC (ISO) Agar contains 0.1g per litre chloramphenicol in the agar base. This removes the need for supplementation and handling of the chloramphenicol.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
Specifications
Specifications
| Content And Storage | 10°C to 30°C |
| Description | Pre-supplemented Dichloran Rose-bengal Chloramphenicol (DRBC) (ISO) Agar (Dehydrated) |
| Form | Powder |
| Product Type | Agar |
| Quantity | 500 g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.