Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pre-supplemented Dichloran Rose-bengal Chloramphenicol (DRBC) (ISO) Agar (Dehydrated)

Catalog No. p-7079181
Click to view available options
Quantity:
500 g
2.5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Isolate and enumerate yeast and molds associated with food spoilage with this pre-supplemented DRBC medium while inhibiting non target bacteria.

Isolate and enumerate yeasts and molds associated with food spoilage with Thermo Scientific™ Oxoid™ Pre-supplemented Dichloran Rose-Bengal Chloramphenicol (DRBC) (ISO) Agar Base (Dehydrated) while adhering to ISO 21527-1: 2008. Due to the presence of antibiotic Chloramphenicol in the formulation, the medium provides selectivity against bacteria as well as reducing harmful chemical handling in the laboratory.

Use Pre-supplemented Dichloran Rose-Bengal Chloramphenicol (DRBC) Agar Base for selective isolation and enumeration of viable yeast and molds in food products with a water activity above 0.95 aW.
  • Adheres to ISO 21527-1: 2008 standards.
  • Selective for viable yeast and mold samples in food samples.
DRBC (ISO) is a modification of the original Rose-Bengal Chloramphenicol Medium1 and differs as the Rose-Bengal content has been optimized by adding Dichloran and by changing the pH to 5.6.

The inclusion of chloramphenicol in this medium suppresses bacterial growth, allowing molds, and yeasts to grow. The inhibition of spreading molds and the restriction of their colony size results in improved enumeration and detection. The reduced pH of DRBC (ISO) Agar increases the inhibition of yeasts by the rose-bengal. Pre-supplemented DRBC (ISO) Agar contains 0.1g per litre chloramphenicol in the agar base. This removes the need for supplementation and handling of the chloramphenicol.

Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 10°C to 30°C
Description Pre-supplemented Dichloran Rose-bengal Chloramphenicol (DRBC) (ISO) Agar (Dehydrated)
Form Powder
Product Type Agar
Quantity 2.5 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.