Learn More
Valutek™ Pre-Wetted Pressure Heated Polyester Wiper
Description
- The pre-wetted format enhances the wipers absorbent capacity, effectiveness, and reduces airborne contamination from traditional solvent bottle dispensers, spray and spills
- 100% continuous-filament double-knit polyester fiber
- Pressure-heat sealed edge for lowest available fiber contamination
- Pre-wetted with specified IPA/DI Water concentrations to a 38% saturation level
- The pre-wet solution increases both cleaning efficiency and cleaning protocol consistency
- Very low levels of particles, fibers, ions and extractables
- All Valutek™ pre-wetted wipers are assembled in a Class 10 cleanroom
under Valutek™ inspection and strict process control to ensure Valutek™ quality standard and product specifications - This product is recommended for use in a cleanroom Class 1 -10 (ISO 3-4) critical environment
- It is designed for use in wet cleaning of critical surfaces where control of flammable solvents and flammable solvent concentrations is required
- Various IPA blend levels are ideal for removing different cleaning and disinfecting residues in regulated environments
Specifications
Specifications
| Dimensions (L x W) | 9 x 9 in. |
| Packaging Quantity | 150 /Pail, 2 Pails/Cs. |
| Saturation Solution | 70% IPA/30% DI H2O |
| Sterility | Non-sterile |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.