Learn More
Bio Rad Laboratories Precision Plus Protein™ Dual Color Standards, 500 µl
Supplier:  Bio Rad Laboratories 1610374
Encompass
500 μl, mixture of 10 recombinant proteins (10–250 kD), 8 blue-stained bands, and 2 pink reference bands (25 and 75 kD), 50 applications
Catalog No. NC9965091
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.