Learn More
Cayman Chemical Precelys 2ml GrIndI 2box
SDP
Supplier:  Cayman Chemical 179522 BOX
Encompass
A mixed antagonist at dopamine D2 D3 and serotonin 5-HT2A receptors (Kis 0.14 0.49 and 0.81 nM respectively) that also binds to 5-HT6 (Ki 11.7 nM) developed as a novel antipsychotic that displays 20- and 94-fold higher affinity for D2 receptors compared to haloperidol and risperidone respectively
Catalog No. 50-296-5155
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.