Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ Precision Balance ME603T/00

Catalog No. 01670591 Shop All METTLER TOLEDO Products
Change view
Click to view available options
:
Precision Balance ME603T/00; Capacity: 620 g; Diameter (English) Outer Pan: 4.7 in.; Dimensions (D x W x H): 319 x 210 x 289 mm (12.6 x 8.3 x 11.4 in.); Readability: 1 mg; Stabilization Time: 1.5 s
Catalog No.
01-670-591 Precision Balance ME603T/00; Capacity: 620 g; Diameter (English) Outer Pan: 4.7 in.; Dimensions (D x W x H): 319 x 210 x 289 mm (12.6 x 8.3 x 11.4 in.); Readability: 1 mg; Stabilization Time: 1.5 s
1 options
This item is not returnable. View return policy

Catalog No. 01-670-591

Supplier: Mettler Toledo™ 30704481

This item is not returnable. View return policy

Mettler Toledo™'s Standard ME-T Precision Balances ensure high quality precision weighing, from their large color touchscreens with onscreen guidance to their high quality materials for long-lasting weighing performance.

  • Reliable for simple, fast, and efficient weighing
  • Passcode protection for extra security
  • FACT fully automatic adjustment using balance's internal weights ensure ongoing accurate weighing performance
  • Color touchscreen features an intuitive user interface with simple menus and clear icons
  • Touchscreen is compatible with cloth or rubber gloves
  • Metal base, stainless steel weighing pan and overload protection ensure reliable performance, even under intense daily use
TRUSTED_SUSTAINABILITY

Specifications

Automation Interface USB-B (To Device), RS232 (Integrated/Optional), USB-A (To Device), Bluetooth (Optional)
Capacity 620 g
Display Type Color Touchscreen
Readability 1 mg
Material ABS, Die-cast Aluminum
Pan Shape Round
Repeatability 0.7 mg
Stabilization Time 1.5 sec.
Type Laboratory Balance
Dimensions (D x W x H) 319 x 210 x 289 mm (12.6 x 8.3 x 11.4 in.)
Diameter (English) Outer Pan 4.7 in.
Diameter (Metric) Outer Pan 120 mm
Legal for Trade No
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.