Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Precision Boyle's Law Apparatus
SDPSCI_ED

Catalog No. S27836 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Includes:
Bicycle Pump, Activity Guide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S27836 Bicycle Pump, Activity Guide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27836 Supplier United Scientific Supplies Supplier No. PBLA01
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Quickly and easily perform a quantitative verification of Boyle's Law

  • Apparatus yields more reliable results than standard syringe-based devices
  • Precisely control pressure with included bicycle pump
  • Dial gauge shows both kPa and psi indicators
  • Built-in length scale features movable markers to assist with measurement
  • Activity guide included

Specifications

For Use With (Application) Quantitative Verification of Boyle's Law
Product Type Boyle's Law Apparatus
Includes Bicycle Pump, Activity Guide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.