Learn More
PDC Healthcare Ident-Alert™ Color Coded Snaps
Description
- Use with snap closure wristbands to quickly and cost-effectively identify special patient conditions
- Provide convenience by allowing caregivers to classify multiple patient conditions
- Soft, flexible material with rounded edges for patient comfort
- Enhance patient safety with large, easy-to-read text that identifies at-risk patients
- Locking SecurSnap™ safety feature prevents transfer of alert to another patient
- Ident-Alert™ Snap can be removed without destroying the admitting wristband
Specifications
Specifications
| Color | Red |
| For Use With (Application) | Snap closure wristbands |
| Message | Allergy |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.