Learn More
United Scientific™ Precision Fresnel Biprism Interface Apparatus
SDPSCI_ED
Catalog No. S23931 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
:
Fresnel Biprism Interface
Catalog No.
S23931 Fresnel Biprism Interface
1 options
Catalog No. S23931 Supplier United Scientific™ Supplier No. PFB001
Precision optics allow fringe position measurements to 1/100 mm. The Precision Fresnel Biprism Interface Apparatus offers easy alignment and adjustment of the two virtual light sources.
  • Perform Fresnel's Biprism Experiment on a 1 m optical bench.
  • Precision optics allow fringe position measurements to 1/100 mm.
  • Easy alignment and adjustment of the two virtual light sources.
  • Fresnel's Biprism Experiment is the fundamental example of common path interference by division of wavefront.
  • Light from a slit passes through side-by-side prisms with very shallow angles to separate it into two almost parallel beams.
  • When these beams are brought to overlapping by a convex lens, they interact as if they originated from two virtual sources side by side and generate equally-spaced fringes.
  • The original version of the experiment by Fresnel is difficult to perform because it requires precise adjustment of the slit and prisms, a dark room, and a large amount of space to separate the fringes for easy viewing.
Specifications
Product Type Optics Set
Shape Various