Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Precision ID DL8 Kit
Specifications
Specifications
| Content And Storage | • 4 cartridges Ion AmpliSeq Chef reagents DL8, store at -5 to -30°C • 4 cartridges Ion AmpliSeq Chef solutions DL8, shipped at room temperature, store at 2–8°C upon receipt • 4 boxes Ion AmpliSeq supplies (each box contains: Ion AmpliSeq tip cartridge L8, framed PCR foil seal, enrichment cartridge), store at room temperature • 1 set of 4 IonCode in PCR plates (the set includes: IonCode 0101-0108 in 96-well PCR plate (red), IonCode 0109-0116 in 96-well PCR plate (yellow), IonCode 0117-0124 in 96-well PCR plate (green), IonCode 0125-0132 in 96-well PCR plate (blue)), store at room temperature |
| Format | 96-well Plate |
| Product Type | Precision ID DL8 Kit |
| Quantity | 32 reactions |
| Sequencing Type | Genome & DNA Sequencing |
| Shipping Condition | Dry Ice |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?