Learn More
Eisco™ Precision pH Test Strips - Narrow Range 3-6 pH, Quick and Accurate Testing in 0.5 pH Increments
SCI_ED
Catalog No. S02931 Shop All Eisco Products
Change view
Click to view available options
pH Range:
3-6 pH
Catalog No. pH Range
S02931 3-6 pH
1 options
Catalog No. S02931 Supplier Eisco™ Supplier No. CH202404
Add to Cart
Edge
Add to Cart
Eisco Labs Precision pH test strips offer quick and accurate testing within a narrow 3-6 pH range, measuring in 0.5 pH increments. Ideal for assessing mildly acidic to neutral substances.
  • High precision: Accurately measures pH levels from 3 to 6 in 0.5 increments, allowing for detailed pH monitoring.
  • Formulated for foods: Ideal for testing mildly acidic to neutral pH substances like sushi rice and salsa.
  • Easy to use: Simply dip one end of the strip into your sample for 1-2 seconds without touching the test pad, ensuring precise results.
  • Instant reading: Remove, shake off excess liquid, and immediately compare with the color chart for quick pH measurements.
  • Secure handling: Each strip is designed for direct testing without finger contact on the sensitive pad, maintaining strip integrity.
  • Optimize your pH testing with our Precision pH Test Strips, designed to measure pH levels from 3 to 6 in detailed 0.5 increments.
  • These strips are simple to use: immerse one end briefly, remove, and immediately compare the color with the included chart for a quick assessment.
  • The design ensures controlled handling, preventing contact with the sensitive test pad for consistent performance.
  • Perfect for various testing needs, these strips provide clear, rapid results without the hassle.
  • They are an essential tool for anyone needing reliable pH measurements in a variety of contexts.
Specifications
pH Range 3-6 pH
Type pH 3-6 Test Strips