Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Precocene II, 99%, Thermo Scientific™

Catalog No. AC187032500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC187032500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC187032500 Supplier Thermo Scientific Chemicals Supplier No. 187032500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 644-06-4
Molecular Formula C13H16O3
Molecular Weight (g/mol) 220.268
InChI Key PTIDGSWTMLSGAH-UHFFFAOYSA-N
Synonym precocene ii, ageratochromene, precocene 2, pricocene ii, 6,7-dimethoxy-2,2-dimethyl-2h-chromene, precocen 2, 2h-1-benzopyran, 6,7-dimethoxy-2,2-dimethyl, unii-1w2r4try5h, 6,7-dimethoxy-2,2-dimethyl2h-1-benzopyran, 6,7-dimethoxy-2,2-dimethyl-2h-benzo b pyran
PubChem CID 12565
ChEBI CHEBI:35606
IUPAC Name 6,7-dimethoxy-2,2-dimethylchromene
SMILES CC1(C=CC2=CC(=C(C=C2O1)OC)OC)C

Specifications

Melting Point 44°C to 47°C
Color Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Quantity 250 mg
Merck Index 14, 7718
Formula Weight 220.27
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material Precocene II, 99%
EINECSNumber 211-408-5
RTECSNumber DJ2527000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.