Learn More
Cole-Parmer Idex Biocompatible Precolumn Filter, HPLC, PEEK/PCTFE, 0.5 µm, 10-32 Coned
Supplier:  Cole-Parmer 0201035
Encompass
Precolumn filters offer secure protection for analytical columnsEconomical protection for larger columns and injectionsFeature 10-32 coned ports for connection to 1/16" OD tubingReplaceable stainless steel (SS) and PEEK® fritsMax pressure 5000 psi (345 bar)
Catalog No. NC2818750
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.