Learn More
Medchemexpress LLC HY-B1832 1g Medchemexpress, Prednisone acetate CAS:125-10-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB18321G
Encompass_Preferred
Medchemexpress, HY-B1832 1g Prednisone acetate CAS:125-10-0 Prednisone acetate (Prednisone 21-acetate), the acetate salt form of prednisolone, is a glucocorticoid receptor agonist with anti-inflammatory and immunomodulating properties [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3017
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.