Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prednisone, Anhydrous, Micronized, USP, 97-102%, Spectrum™ Chemical
SDP

Catalog No. 18605804 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-804 100 g Amber Glass Bottle
18-605-802 5 g Amber Glass Bottle
18-605-803 25 g Amber Glass Bottle
18-605-807 500 g Amber Glass Bottle
18-605-805 1 kg Amber Glass Bottle
18-605-806 5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-605-804 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. PR103100GM
Only null left
Add to Cart
Add to Cart

PR103, 53-03-2, C21H26O5

Spectrum™ Chemical Prednisone, Anhydrous, Micronized, USP is a steroid medication used to treat certain types of allergies, inflammatory conditions, autoimmune disorders, and cancers. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 53-03-2
Molecular Formula C21H26O5
Molecular Weight (g/mol) 358.43
InChI Key XOFYZVNMUHMLCC-ZPOLXVRWSA-N
IUPAC Name (1R,3aS,3bS,9aR,9bS,11aS)-1-hydroxy-1-(2-hydroxyacetyl)-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthrene-7,10-dione
SMILES C[C@]12CC(=O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO

Specifications

CAS Max % 1
Ignition Residue Negligible
Melting Range 230-235°C
Packaging Amber Glass Bottle
Quantity 100 g
Specific Rotation +167° to +175°
Percent Purity 97 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Prednisone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.