Description
- Cleaner - Effective disinfectant as well as a cleaner resulting in added confidence that disinfection can occur
- Faster - Rapid and realistic contact times on a broad spectrum of difficult to kill pathogens
- Responsible - Users are not exposed to VOCs (volatile organic compounds) and PPE is not required as per health and safety rating
- Utilizes AHP™, a patented synergistic blend of commonly used, safe ingredients that when combined with low levels of hydrogen peroxide dramatically increase its potency and cleaning performance
- 5 Minutes Bactericidal, Virucidal, Tuberculocidal, Fungicidal (1:16)
- 3 minute Broad-Spectrum Sanitizing (1:128)
- 30 second Broad-Spectrum Sanitizing (1:16)
Specifications
Specifications
| Product Type | Concentrated Disinfectant Solution |
| Container Type | Drum |
| Volume (English) | 55 gal. |
| Volume (Metric) | 200 L |