Learn More
PIP Prefilled Mega T-Fit™ Ear Plug Dispenser
Catalog No. 17100839 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Noise Reduction Rating (NRR):
32
Catalog No. Noise Reduction Rating (NRR)
17-100-839 32
1 options
Catalog No. 17-100-839 Supplier PIP Supplier No. 267HPD510500
Add to Cart
Add to Cart
Table Top or Wall Mount Dispenser. Twist to dispense. Holds 500 pair of 267-HPF510. Transparent Top. Uses Refill Packs. Earplugs Included.
  • Prefilled dispenser includes 500 pairs of PIP Mega T-Fit ear plugs
  • Table top install and easy to use
  • Controlled release mechanism ensures the correct number of plugs are dispensed into a collection tray
  • Sufficient capacity to supply large production areas
  • Transparent design to monitor the number of earplugs remaining
Specifications
ANSI S3.1974 Yes
CSA Z94.2 Class AL
Size Universal
High Visibility Yes
Noise Reduction Rating (NRR) 32
Corded Uncorded
Plug Shape T-Shape
Product Type Ear Plugs
Disposable Single-use
Metal Detectable No
Dielectric No
EN 352.2002 EN 352-2