Learn More
Lee Biosolutions Inc Pregnant Mare Serum Gonadotropin (PMSG) | 2.5 kIU
Supplier:  Lee Biosolutions Inc 493102.5
Encompass_Preferred
Pregnant mare serum gonadotropin (PMSG) is a glycoprotein hormone synthesized in the domestic horse placenta. Commercial preparations are isolated from the serum of pregnant mares. The hormone is a dimer of non-covalently associated alpha and beta subunits. It is a unique member of the gonadotropin family in that it contains high levels of FSH and LH activity. Its ability to stimulate ovulation is determined in a bioassay, and the results are expressed as international units (IU). PMSG that has been purified to homogeneity has approximately 16,000 IU/mg protein. PMSG may be used to induce ovulation in livestock prior to artificial insemination.
Catalog No. NC1663485
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.