Learn More
Abcam Pregnenolone-16alpha-carbonitrile, Cytochrome P-450 Inducer, Solid, >99% Purity, 100 mg
Description
- Molecular Weight: 341.5 Da
- Purity: >99%
- Form: Solid
- Mechanism of Action: Cytochrome P-450 inducer; activates PCN1 transcription
- Receptor Ligand: Xenobiotic receptor (SXR)/pregane X receptor (PXR) ligand
- Applications: Active in vitro and in vivo
- Storage: Store at -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Pregnane X Receptor (PXR), Steroid and Xenobiotic Receptor (SXR) |
| Shipping Condition | Ambient |
| Form | Solid |
| Content And Storage | Store at -20°C. |
| Quantity | 100 mg |
| For Use With (Application) | Cytochrome P-450 induction, PCN1 transcription activation, SXR/PXR ligand research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.