Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Pregnenolone
SDP

Catalog No. 14186132 Shop All Medisca Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-186-132 1 kg
14-185-564 100 g
14-185-565 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-186-132 Supplier Medisca Supplier No. 076909
Add to Cart
Add to Cart

Pregnenolone is a white or almost white crystalline powder.

Pregnenolone is a white or almost white crystalline powder.

Specifications

Chemical Name or Material Pregnenolone
Packaging Jar 2500ml, White, HDPE, Round, 89-400, TRIVENI-DB, CT
CAS 145-13-1
Type Drug Standard
Molecular Weight (g/mol) 316.47
Quantity 1 kg
Percent Purity ≥99.0 %
Grade Ungraded
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.