Learn More
Specifications
Specifications
| CAS Max % | 1 |
| Boiling Point | 443.26°C (829.87°F) |
| Loss on Drying | 0.005 |
| Melting Range | 183° to 194°C |
| Packaging | Amber Glass Bottle |
| Quantity | 25 kg |
| Specific Rotation | +27° to +30° |
| Molecular Weight (g/mol) | 316.47 |
| Percent Purity | 0.98 |
| Grade | Reagent |
| Show More |
CAUTION: For use by qualified personnel for research and development or other authorized use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
