Learn More
Apexbio Technology LLC Preladenant, 10mg. Cas: 377727-87-2 MFCD: MFCD17167056. Adenosine A2A receptor antagonist
SDP
Supplier:  Apexbio Technology LLC A373510
Encompass_Preferred
Preladenant is a high selective antagonist of adenosine A2A receptor with Ki value of 1.1 nM [1].Parkinson's Disease is characterized by the loss of dopaminergic neuronal projection. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4832
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.