Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRELID1 Monoclonal Antibody (OTI2H5), TrueMAB™, OriGene
SDP

Catalog No. 501683745
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-3745 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-3745 Supplier OriGene Supplier No. TA810978
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

May be important for the development of vital and immunocompetent organs.

Specifications

Antigen PRELID1
Applications Western Blot
Classification Monoclonal
Clone OTI2H5
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PRELID1
Gene Accession No. Q9Y255
Gene Alias CGI-106, PRELI, PX19, SBBI12
Gene Symbols PRELID1
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 36-219 of human PRELID1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27166
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.