Learn More
ABS Premier Pharmacy/Vaccine Built-in Undercounter Freezer
GREENER_CHOICESDP
Catalog No. 22651531 Shop All Nor-Lake Scientific Products
Change view
Click to view available options
Capacity (English):
4.2 cu. ft.
Catalog No. Capacity (English)
22-651-531 4.2 cu. ft.
1 options
This item is not returnable. View return policy
Catalog No. 22-651-531 Supplier ABS Supplier No. PHABTHCUCBI0420A
Add to Cart
Add to Cart
This item is not returnable. View return policy
Enjoy the compact design and professional cooling power of ABS built-in undercounter freezers. These units are designed specifically for pharmacies and medical settings.
Experience a range of innovative features to enhance your work.
  • Microprocessor temperature control
  • Array of temperature alarms
  • Keyed door locks
  • Probe access
  • Natural hydrocarbon refrigerants
Built-in units are designed with front ventilation systems, allowing easy installation into a bench or countertop. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency. Note: Some models are available with auto defrost and ADA compliance.
Specifications
Type Freezer
Capacity (English) 4.2 cu. ft.
Capacity (Metric) 119 L
Temperature Range -15°C to -25°C
Door Style Solid Swing
No. of Doors 1
Cabinet Material Scratch-resistant Painted Steel
Exterior Finish Powder-coated Steel
Shelves 3
Certifications/Compliance cULus
Style Undercounter
Monitoring Options Alarms, Remote Alarm Contacts
Plug Type NEMA 5-15
Voltage 115 V
Compressor H-drive
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov