Learn More
ABS Premier Pharmacy/Vaccine Built-in Undercounter Refrigerator
GREENER_CHOICESDP
Catalog No. 22651522 Shop All Nor-Lake Scientific Products
Change view
Click to view available options
Capacity (English):
4.6 cu. ft.
Catalog No. Capacity (English)
22-651-522 4.6 cu. ft.
1 options
This item is not returnable. View return policy
Catalog No. 22-651-522 Supplier ABS Supplier No. PHABTHCUCBI0404
Add to Cart
Add to Cart
This item is not returnable. View return policy
Enjoy the compact design and professional cooling power of ABS built-in undercounter refrigerators. These units are designed specifically for pharmacies and medical settings.
Experience a range of innovative features to enhance your work: - Microprocessor temperature control - Array of temperature alarms - Keyed door locks - Probe access - Natural hydrocarbon refrigerants Built-in units are designed with front ventilation systems, allowing easy installation into a bench or countertop. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency. Note: Some models are available with ADA compliance.
Specifications
Type Refrigerator
Capacity (English) 4.6 cu. ft.
Capacity (Metric) 130 L
Temperature Range 2°C to 8°C
Door Style Solid Swing
No. of Doors 1
Shelves 3
Cabinet Material Stainless Steel
Exterior Finish Powder-coated Steel
Certifications/Compliance cULus, ENERGYSTAR
Style Undercounter
Monitoring Options Alarms, Remote Alarm Contacts
Compressor H-drive
Plug Type NEMA 5-15
Voltage 115 V
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov