Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Premium Universal Swivel Boss Head Clamp
SCI_ED

Catalog No. S13922 Shop All Eisco Products
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S13922 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S13922 Supplier Eisco™ Supplier No. PRBH04
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Premium Universal Swivel Boss Head Clamp adjusts to any angle (dual 360 rotation) and accepts rods of up to 16 mm diameter.

  • Threaded metal one piece clamping screws
  • Able to hold rods up to 16 mm in diameter
  • Both bossheads can swivel 360 degrees
  • Made of high strength powder coated alloy
  • All metal constructions to carry heavy loads and prevent torque damage

Specifications

Autoclavable Autoclavable
Type Swivel Clamp
Material Nickel-plated
Holds Rods up to 16 mm diameter
For Use With (Application) Adjusts to any angle, Dual 360 rotation
Color Blue
Jaw Type Swivel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.