Learn More
Columbia Cleanroom Premium Cleanroom Paper, Latex - Free
Description
- The same high-quality paper found in all our Class 10 cleanroom notebooks.
- This is the best cleanroom sheet available.
- Our latex-free Premium cleanroom paper is a cut above our traditional latex-free cleanroom paper.
- Its purpose is simple: if your company is seeking the very best in cleanroom paper quality, whether for copying, writing, monthly cleanroom reports, or lab records, you will not find a better sheet.
- With its rich, natural finish, acid-free compound, and absence of chlorine, our Class 10, latex-free, Premium sheet is archival quality.
- Our latex-free Premium sheet is Forest Stewardship Council (FSC) certified.
- We offer latex-free Premium in 22 and 28 pound, 8.5 x 11 in. and 11 x17 in.
- 100% made in the USA.
- ISO 3 certified.
- The lowest shed rate when torn.
- Superior smoothness.
- No bleeding from pen ink or copier toner.
- Purpose-made for controlled environments.
- Ideal for copiers, laser, and inkjet printers.
- Distinct quality and superior finish.
- Unlike any other cleanroom sheet on the market.
- 1250 sheets per case.
Specifications
Specifications
| Material | Paper |
| Color | White |
| Dimensions (L x W) | 11 x 17 in. |
| Product Type | Paper |
| Weight (Metric) | 9.97 kg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.