Learn More
Eisco™ Premium Insect Entomology Dissection Pins
SCI_ED
Catalog No. S24851 Shop All Eisco Products
Change view
Click to view available options
Quantity:
100/Pk.
Catalog No. Quantity
S24851 100/Pk.
1 options
Catalog No. S24851 Supplier Eisco™ Supplier No. DISPINS00
Add to Cart
Add to Cart
With black enamaled coating for rust protection.
  • Made from spring steel with nylon gold heads
  • Museum grade
Specifications
Product Type Premium Insect Entomology Dissection Pin
Autoclavable Not Autoclavable
Color Black
Material Spring Steel with Nylon Gold Heads
Includes Double black-enamel coating
Quantity 100/Pk.
Shape Size 00
Length (English) 1.7 in.
Length (Metric) 4.31 cm