Learn More
Eisco™ Premium Large Three Finger Clamp w/ Boss head
SCI_ED
Catalog No. S13914 Shop All Eisco Products
Change view
Click to view available options
Color:
Blue
Catalog No. Color
S13914 Blue
1 options
Catalog No. S13914 Supplier Eisco™ Supplier No. PRCMP07BH
Add to Cart
Add to Cart
Choose durable lab equipment that will last for years. The EISCO Premium Large Three Finger Clamp w/ Boss head is an industrial clamp that has rubber coated fingers.
  • Clamp constructed of powder coated steel, 3 finger rubber coated clamp attached to aluminum rod
  • Maximum clamp opening is 8.5 cm
  • 14 cm connecting rod
  • Completely metal constructions, thread fitted actuating joints
  • 10 mm diameter connecting rod
Specifications
Autoclavable Autoclavable
Type 3-Prong Extension Clamp
Material Zinc Die Cast, Powder Coated
Color Blue
Jaw Type 3-Prong
Coating Rubber