Description
- Available Sizes: Small, Medium, Large, Narrow
- Narrow, tapered blades ideal for patients with smaller introitus or vaginal stenosis
- Smooth screw-lock mechanism for precise and stable opening
- Rounded, polished edges enhance patient comfort during exams
- Crafted from medical-grade stainless steel—durable, corrosion-resistant, and fully autoclavable
- Suitable for OB/GYN, primary care, emergency medicine, and outpatient procedures
Specifications
Specifications
| Product Type | Vaginal Speculum |
| Grade | Medical |
| Size | Narrow |
| Autoclavable | Autoclavable |