Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Premium Safety Needle System - Tube Touch

Catalog No. 22040361
Change view
Click to view available options
Size:
20 G
22 G
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
22-040-361 20 G
22-040-363 22 G
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-040-361 Supplier Greiner Bio-One Supplier No. 450255
This item is not returnable. View return policy

Manual activation of the safety device not needed

A passive safety engineered device (SED), fully protects the individual against needlestick injury at the time the venipuncture is terminated or interrupted.

  • Activation via tube insertion
  • Features automatic safety shield
  • Position-guide on safety shield aids skin penetration visualization
  • No blood splatter
  • Color-coded cap for easy needle gauge identification
  • Ergonomically designed with recessed thumb placement
  • Activation and venipuncture termination simultaneously
  • Cut away, flanged holder for low angle of insertion

  • Simple and eco-friendly packaging
  • Perforated device labeling

TRUSTED_SUSTAINABILITY

Specifications

Size 20 G
Length (Metric) Needle 31.75 mm
Sterility Sterile
Length (English) Needle 1.25 in.
Product Type Safety Needle System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.