Learn More
Eisco™ Premium Three Finger Clamp with Dual Screw Adjustments PROMO
SCI_ED
Three Finger Clamp with Dual Screw Adjustments is an industrial three finger, rubber-coated extension lab clamp with dual screw adjustments.
Supplier:  Eisco™ PRCMP06PROMO
Catalog No. S13911W25
This item is not returnable. View return policy
Description
Description
  • Heavy duty spring loaded clamp jaw with dual screw adjustment to fine tune any fitting
  • Made of steel and powder coated alloy with all metal adjustment screws
  • 3 rubber coated clamp fingers
  • 6 cm maximum opening clamp
  • 12.5 cm steel connecting rod
Specifications
Specifications
Clamp
6 cm max. opening
3-Prong
Steel
Gray
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.