Learn More
Eisco™ Premium Three Finger Clamp
SCI_ED
Catalog No. S13917 Shop All Eisco Products
Change view
Click to view available options
Color:
Blue
Catalog No. Color
S13917 Blue
1 options
Catalog No. S13917 Supplier Eisco™ Supplier No. PRCMP09
Add to Cart
Add to Cart
Choose durable lab equipment that will last for years. The Premium Three Finger Clamp is an industrial three finger, cork-lined extension clamp.
  • Clamp constructed of powder coated steel
  • Maximum clamp opening is 8.5 cm
  • 14 cm connecting rod
  • Completely metal constructions, thread fitted actuating joints
  • 10 mm diameter connecting rod
Specifications
Autoclavable Not Autoclavable
Type 3-Prong Clamp
Material Zinc Die Cast, Plastic Coated
Color Blue
Jaw Type 3-Prong
Coating Cork