Learn More
Eisco™ Premium Two Prong Clamp w/ Dual Screw Adjustments
SCI_ED
Catalog No. S13907 Shop All Eisco Products
Change view
Click to view available options
Color:
Blue
Catalog No. Color
S13907 Blue
1 options
Catalog No. S13907 Supplier Eisco™ Supplier No. PRCMP04
Add to Cart
Add to Cart
Choose durable lab equipment that will last for years. The EISCO Premium Two Prong Clamp w/ Dual Screw Adjustments is an industrial, rubber-coated lab clamp with dual screw adjustments.
  • Heavy duty spring loaded clamp jaw with dual screw adjustment to fine tune any fitting
  • Made of steel and powder coated alloy with all metal adjustment screws
  • 2 rubber coated clamp tongs
  • 7 cm maximum clamp opening
  • 12.5 cm connection rod
Specifications
Autoclavable Autoclavable
Type Adjustable Clamp
Material Zinc Die Cast, Powder Coated
Color Blue
Jaw Type Dual Adjustment
Coating Vinyl