Learn More
Microsolv Technology Corporation Prep HPLC Column, HPS Silica, 5um, 120A. 21.2mm ID x 250mm Length. 1 EA.
Supplier:  Microsolv Technology Corporation 75001P250
Encompass_Preferred
This Cogent HPS Silica™ Stationary Phase has been a premium value and used by HPLC laboratories for many years. This proprietary Silica is not modified in any way and is ideal for purification of polar compounds by preparative HPLC. With a 5 µm Particle, this column works well with higher flow rates and lower back pressures compared with smaller particles and is often used for hydrophilic or polar compounds in Normal Phase HPLC. Compare its performance to Inertsil and Luna products. A specialized HPLC System is required for columns this size.
Catalog No. 50-227-1047
Only null left
Add to Cart
Request a Quote
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.