Learn More
Microsolv Technology Corporation Prep HPLC Column, UDC Cholesterol, 4um, 100A. 21.2mm ID x 150mm Length, Preparative. 1 EA.
Supplier:  Microsolv Technology Corporation 69069P21150
Encompass_Preferred
This UDC Cholesterol™ HPLC is made with Cogent TYPE-C™ Silica , and provides unique Selectivity due to different surface chemistry, compared to typical Reversed Phase Columns. This Silica Hydride Phase is modified with an 11carbon chain with a cholesterol molecule attached at the terminus. This gives this column the ability to form “slots” and separate some compounds based on shape. Also, due to the direct silicon carbon bonds, this column has no need for end-capping. Use in Reversed Phase, Normal Phase and Aqueous Normal Phase (ANP). Chosen for analyzing or collecting steroidal and other hard to retain compounds including some lipids, peptides, proteins & oligonucleotides. Faster Equilibration than other Reversed Phase or HILIC Columns Very Hydrophobic Robust Column Lifetimes Temperature and Solvent Changes Selectivity A specialized HPLC System is required for columns this size.
Catalog No. 50-227-0902
Only null left
Add to Cart
Request a Quote
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.