Learn More
Microsolv Technology Corporation Prep Guard Column Cartridge, RP C18 Wide Pore, 10um, 300A. 30mm ID x 10mm Length. 1 EA.
Supplier:  Microsolv Technology Corporation 68018P30PG13
Encompass_Preferred
This Guard Column Cartridge is packed with 300Å, 10µm Cogent RP C18™ Stationary Phase. It is double end capped ( fully ), with traditional bonding of an 18 carbon silane to the highest purity silica available. It is designed to protect your RP C18 Wide Pore, Prep Column from particulate and contaminants that will adsorb to it. Packed in a Stainless Steel Tube, it has SS Frits. Use this Guard Column Cartridge with a 30mm ID Prep Column Holder.
Catalog No. 50-227-0194