Learn More
Eisco™ Cerebellum Section (Mammal), HE Stain
SCI_ED
Catalog No. S30379 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1
Catalog No. Quantity
S30379 1
1 options
Catalog No. S30379 Supplier Eisco™ Supplier No. BS18236
Add to Cart
Add to Cart
Prepared microscope slide of a section of mammalian cerebellum. Stained with hematoxylin and eosin for better visualization of characteristic tissues and structures. Great choice for biology classrooms studying nervous system.
  • pre-cleaned glass slide with ground edges
  • size 1x3 in. (25x77 mm)
  • professionally stained samples for best visualization
  • great choice for biology classrooms to explore structure-function relationships as per NGSS standards
  • showing characteristic tissues and structures
Specifications
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Quantity 1
Specimen Type Mammal Cerebellum
Stain Type H and E stain
Size 1 x 3 in.
Slide Type Prepared Slide